2-(Benzyloxy)-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15249-1G 15249-500MG 15249-5G 15249-25G
77465-076EA 221.45 USD
77465-076 77477-844 77487-982 77498-674
2-(Benzyloxy)-4-(trifluoromethyl)benzaldehyde ≥97%
2-(Benzyloxy)-4-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₁F₃O₂
MW: 280.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26395940
CAS Number: 873194-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR