4-Trifluoromethyl-benzoic acid 2,2-dimethyl-propyl ester 95

Supplier: AOBChem USA

Warning

15188-1G 15188-500MG 15188-5G 15188-25G
77465-068EA 138.02 USD
77465-068 77477-836 77487-974 77498-664
4-Trifluoromethyl-benzoic acid 2,2-dimethyl-propyl ester 95
4-Trifluoromethyl-benzoic acid 2,2-dimethyl-propyl ester

Precaución: For research use only

Formula: C₁₃H₁₅F₃O₂
MW: 260.252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976796
CAS Number: 204779-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR