4-Bromo-2-iodobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

151304-1G 151304-500MG 151304-5G
77465-060EA 577.83 USD
77465-060 77477-828 77487-964
4-Bromo-2-iodobenzo[b]thiophene ≥95%
4-Bromo-2-iodobenzo[b]thiophene

Precaución: For research use only

Formula: C₈H₄BrIS
MW: 338.991 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619342
CAS Number: 2181829-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR