(4-Bromobenzo[b]thiophen-2-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

151300-1G 151300-500MG 151300-5G
77465-056EA 975.41 USD
77465-056 77477-824 77487-960
(4-Bromobenzo[b]thiophen-2-yl)boronic acid ≥95%
(4-Bromobenzo[b]thiophen-2-yl)boronic acid

Precaución: For research use only

Formula: C₈H₆BBrO₂S
MW: 256.912 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29045013
CAS Number: 2374946-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR