(2,4-Difluoro-3-(trifluoromethyl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

15105-1G 15105-500MG 15105-5G 15105-25G
77465-052EA 256.47 USD
77465-052 77477-820 77487-956 77498-648
(2,4-Difluoro-3-(trifluoromethyl)phenyl)boronic acid ≥97%
(2,4-Difluoro-3-(trifluoromethyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₇H₄BF₅O₂
MW: 225.908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21340321
CAS Number: 769937-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR