2-Chloro-1-(2,3-dichloro-6-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150083-1G 150083-500MG 150083-5G 150083-25G
77465-048EA 415.09 USD
77465-048 77477-816 77487-952 77498-646
2-Chloro-1-(2,3-dichloro-6-fluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,3-dichloro-6-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂Cl₃F₃O
MW: 277.455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841213
CAS Number: 2921712-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR