2-Chloro-1-(2,5-difluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150080-1G 150080-500MG 150080-5G
77465-044EA 594.31 USD
77465-044 77477-812 77487-948
2-Chloro-1-(2,5-difluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,5-difluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₃ClF₄O
MW: 226.555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045709
CAS Number: 1564749-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR