1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150074-1G 150074-500MG 150074-5G 150074-25G
77465-040EA 415.09 USD
77465-040 77477-808 77487-944 77498-642
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃OS
MW: 333.553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841207
CAS Number: 2921896-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR