1-(6-Bromo-2-fluoro-3-propoxyphenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150064-1G 150064-500MG 150064-5G 150064-25G
77465-036EA 415.09 USD
77465-036 77477-804 77487-940 77498-638
1-(6-Bromo-2-fluoro-3-propoxyphenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(6-Bromo-2-fluoro-3-propoxyphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₁H₉BrClF₃O₂
MW: 345.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841197
CAS Number: 2921846-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR