2-Chloro-1-(3,6-dibromo-2-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150059-1G 150059-500MG 150059-5G 150059-25G
77465-032EA 415.09 USD
77465-032 77477-800 77487-936 77498-634
2-Chloro-1-(3,6-dibromo-2-fluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3,6-dibromo-2-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂Br₂ClF₃O
MW: 366.357 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841192
CAS Number: 2921896-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR