1-(2-Bromo-3-chloro-6-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150054-1G 150054-500MG 150054-5G 150054-25G
77465-028EA 415.09 USD
77465-028 77477-796 77487-932 77498-630
1-(2-Bromo-3-chloro-6-fluorophenyl)-2-chloro-2,2-difluoroethanone ≥95%
1-(2-Bromo-3-chloro-6-fluorophenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂BrCl₂F₃O
MW: 321.906 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841187
CAS Number: 2921757-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR