2-Chloro-1-(2,6-difluoro-3-iodophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150045-1G 150045-500MG 150045-5G 150045-25G
77465-024EA 415.09 USD
77465-024 77477-792 77487-928 77498-626
2-Chloro-1-(2,6-difluoro-3-iodophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,6-difluoro-3-iodophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂ClF₄IO
MW: 352.452 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841178
CAS Number: 2921757-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR