2-Chloro-1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150024-1G 150024-500MG 150024-5G 150024-25G
77465-020EA 415.09 USD
77465-020 77477-788 77487-924 77498-622
2-Chloro-1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅Cl₂F₃O
MW: 257.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841158
CAS Number: 2921756-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR