2-Chloro-1-(2,6-difluoro-4-methoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150018-1G 150018-500MG 150018-5G 150018-25G
77465-016EA 415.09 USD
77465-016 77477-784 77487-920 77498-618
2-Chloro-1-(2,6-difluoro-4-methoxyphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,6-difluoro-4-methoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅ClF₄O₂
MW: 256.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841152
CAS Number: 2921752-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR