2-Chloro-1-(5-chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

150001-1G 150001-500MG 150001-5G 150001-25G
77465-012EA 415.09 USD
77465-012 77477-780 77487-916 77498-616
2-Chloro-1-(5-chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(5-chloro-2-fluoro-3-methylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅Cl₂F₃O
MW: 257.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34841136
CAS Number: 2921908-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR