3-Bromo-5-chloro-4-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

149221-1G 149221-500MG 149221-5G 149221-25G
77465-008EA 172.01 USD
77465-008 77477-778 77487-910 77498-610
3-Bromo-5-chloro-4-iodobenzoic acid ≥97%
3-Bromo-5-chloro-4-iodobenzoic acid

Precaución: For research use only

Formula: C₇H₃BrClIO₂
MW: 361.3589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24108573
CAS Number: 1541309-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR