2-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

148919-1G 148919-500MG 148919-5G 148919-25G
77465-004EA 415.09 USD
77465-004 77477-774 77487-904 77498-604
2-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid ≥95%
2-Bromo-4-chloro-5-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₈H₃BrClF₃O₂
MW: 303.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34840412
CAS Number: 2091035-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR