1-Chloro-2-fluoro-3-isopropoxybenzene ≥96%

Supplier: AOBChem USA

Warning

148701-1G 148701-5G 148701-25G
77465-000EA 61.8 USD
77465-000 77487-900 77498-602
1-Chloro-2-fluoro-3-isopropoxybenzene ≥96%
1-Chloro-2-fluoro-3-isopropoxybenzene

Precaución: For research use only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD23114121
CAS Number: 1369791-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR