N-Methyl-5-phenylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

148303-1G 148303-500MG 148303-5G
77464-996EA 546.93 USD
77464-996 77477-768 77487-896
N-Methyl-5-phenylpyridin-2-amine ≥97%
N-Methyl-5-phenylpyridin-2-amine

Precaución: For research use only

Formula: C₁₂H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD20667509
CAS Number: 107351-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR