(3-Chloro-2,6-difluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148071-1G 148071-500MG 148071-5G 148071-25G
77464-992EA 415.09 USD
77464-992 77477-764 77487-892 77498-594
(3-Chloro-2,6-difluorophenyl)(furan-2-yl)methanol ≥95%
(3-Chloro-2,6-difluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₇ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34839774
CAS Number: 2921904-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR