(2,3-Dibromo-6-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148052-1G 148052-500MG 148052-5G 148052-25G
77464-988EA 415.09 USD
77464-988 77477-760 77487-888 77498-590
(2,3-Dibromo-6-fluorophenyl)(furan-2-yl)methanol ≥95%
(2,3-Dibromo-6-fluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₇Br₂FO₂
MW: 349.978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839755
CAS Number: 2921695-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR