(3,6-Dibromo-2-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148050-1G 148050-500MG 148050-5G 148050-25G
77464-984EA 415.09 USD
77464-984 77477-756 77487-884 77498-586
(3,6-Dibromo-2-fluorophenyl)(furan-2-yl)methanol ≥95%
(3,6-Dibromo-2-fluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₇Br₂FO₂
MW: 349.978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839753
CAS Number: 2921848-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR