(3-Bromo-2-chloro-6-fluorophenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148047-1G 148047-500MG 148047-5G 148047-25G
77464-980EA 415.09 USD
77464-980 77477-752 77487-880 77498-582
(3-Bromo-2-chloro-6-fluorophenyl)(furan-2-yl)methanol ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34839750
CAS Number: 2921855-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR