(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

148005-1G 148005-500MG 148005-5G 148005-25G
77464-972EA 415.09 USD
77464-972 77477-744 77487-872 77498-574
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(furan-2-yl)methanol ≥95%
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₇H₁₂BrFO₂
MW: 347.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839710
CAS Number: 2921903-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR