(5-Bromo-2-fluoro-3-(methylthio)phenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

147991-1G 147991-500MG 147991-5G 147991-25G
77464-968EA 415.09 USD
77464-968 77477-740 77487-868 77498-570
(5-Bromo-2-fluoro-3-(methylthio)phenyl)(furan-2-yl)methanol ≥95%
(5-Bromo-2-fluoro-3-(methylthio)phenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀BrFO₂S
MW: 317.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839696
CAS Number: 2921848-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR