2-(4-cyanophenyl)-2-oxoacetic acid ≥96%

Supplier: AOBChem USA

Warning

147884-1G 147884-500MG 147884-5G 147884-25G
77464-964EA 284.28 USD
77464-964 77477-736 77487-864 77498-566
2-(4-cyanophenyl)-2-oxoacetic acid ≥96%
2-(4-cyanophenyl)-2-oxoacetic acid

Precaución: For research use only

Formula: C₉H₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD20639650
CAS Number: 76590-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR