5-Bromo-3,3'-bipyridine ≥96%

Supplier: AOBChem USA

Warning

147699-1G 147699-500MG 147699-5G 147699-25G
77464-960EA 235.87 USD
77464-960 77477-732 77487-860 77498-562
5-Bromo-3,3'-bipyridine ≥96%
5-Bromo-3,3'-bipyridine

Precaución: For research use only

Formula: C₁₀H₇BrN₂
MW: 235.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037813
CAS Number: 15862-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR