3-Bromo-2,6-dichloroaniline ≥97%

Supplier: AOBChem USA

Warning

147418-1G 147418-500MG 147418-5G 147418-25G
77464-956EA 120.51 USD
77464-956 77477-728 77487-856 77498-558
3-Bromo-2,6-dichloroaniline ≥97%
3-Bromo-2,6-dichloroaniline

Precaución: For research use only

Formula: C₆H₄BrCl₂N
Storage Temperature: Ambient
MDL Number: MFCD18432769
CAS Number: 943830-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR