(4-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

147109-1G 147109-500MG 147109-5G 147109-25G
77464-952EA 415.09 USD
77464-952 77477-724 77487-852 77498-556
(4-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane ≥95%
(4-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₄BrFOSi
MW: 277.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34839236
CAS Number: 2921898-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR