5-Bromobenzo[b]thiophene-2-boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1-benzothiophene-2-boronic acid, 2-Borono-5-bromobenzo[b]thiophene

14674-1G 14674-5G 14674-25G
77464-944EA 66.95 USD
77464-944 77487-844 77498-548
5-Bromobenzo[b]thiophene-2-boronic acid ≥95%
5-Bromobenzo[b]thiophene-2-boronic acid

Precaución: For research use only

Formula: C₈H₆BBrO₂S
MW: 256.912 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06801728
CAS Number: 884010-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR