Methyl 2,4-difluoronicotinate ≥95%

Supplier: AOBChem USA

Warning

146640-1G 146640-500MG 146640-5G
77464-940EA 639.63 USD
77464-940 77477-714 77487-840
Methyl 2,4-difluoronicotinate ≥95%
Methyl 2,4-difluoronicotinate

Precaución: For research use only

Formula: C₇H₅F₂NO₂
MW: 173.1169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27922865
CAS Number: 1806304-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR