(E)-1-(2-fluoro-3-(trifluoromethyl)phenyl)but-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

145317-1G 145317-500MG 145317-5G 145317-25G
77464-932EA 415.09 USD
77464-932 77477-708 77487-832 77498-536
(E)-1-(2-fluoro-3-(trifluoromethyl)phenyl)but-2-en-1-ol ≥97%
(E)-1-(2-fluoro-3-(trifluoromethyl)phenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₀F₄O
MW: 234.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34837973
CAS Number: 2148932-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR