(E)-1-(3-Chloro-4-methylphenyl)but-2-en-1-ol ≥96%

Supplier: AOBChem USA

Warning

145312-1G 145312-500MG 145312-5G
77464-928EA 546.93 USD
77464-928 77477-704 77487-828
(E)-1-(3-Chloro-4-methylphenyl)but-2-en-1-ol ≥96%
(E)-1-(3-Chloro-4-methylphenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃ClO
MW: 196.673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30678752
CAS Number: 2154502-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR