(E)-1-(3-(trifluoromethyl)phenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

145305-1G 145305-500MG 145305-5G 145305-25G
77464-924EA 312.09 USD
77464-924 77477-700 77487-824 77498-534
(E)-1-(3-(trifluoromethyl)phenyl)but-2-en-1-ol ≥95%
(E)-1-(3-(trifluoromethyl)phenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₁H₁₁F₃O
MW: 216.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34837969
CAS Number: 2163930-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR