(E)-1-(2-fluorophenyl)but-2-en-1-ol ≥97%

Supplier: AOBChem USA

Warning

145301-1G 145301-500MG 145301-5G
77464-920EA 447.02 USD
77464-920 77477-696 77487-820
(E)-1-(2-fluorophenyl)but-2-en-1-ol ≥97%
(E)-1-(2-fluorophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁FO
MW: 166.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30678612
CAS Number: 1821093-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR