2-Hydroxy-6-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

14529-1G 14529-500MG 14529-5G
77464-912EA 869.32 USD
77464-912 77477-688 77487-812
2-Hydroxy-6-(trifluoromethyl)benzoic acid ≥95%
2-Hydroxy-6-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₈H₅F₃O₃
MW: 206.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848491
CAS Number: 144433-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR