1-Fluoro-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

14511-1G 14511-500MG 14511-5G 14511-25G
77464-908EA 116.39 USD
77464-908 77477-684 77487-808 77498-526
1-Fluoro-2-(methoxymethoxy)benzene ≥95%
1-Fluoro-2-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21195657
CAS Number: 117570-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR