4-Bromo-5-fluoro-2-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

143695-1G 143695-500MG 143695-5G 143695-25G
77464-900EA 398.61 USD
77464-900 77477-676 77487-796 77498-512
4-Bromo-5-fluoro-2-iodobenzoic acid ≥97%
4-Bromo-5-fluoro-2-iodobenzoic acid

Precaución: For research use only

Formula: C₇H₃BrFIO₂
MW: 344.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30297369
CAS Number: 1870155-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR