1,2-Dibromo-3-fluoro-4-(trifluoromethyl)benzene ≥96%

Supplier: AOBChem USA

Warning

143404-1G 143404-500MG
77464-896EA 2544.1 USD
77464-896 77477-672
1,2-Dibromo-3-fluoro-4-(trifluoromethyl)benzene ≥96%
1,2-Dibromo-3-fluoro-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₇H₂Br₂F₄
MW: 321.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784689
CAS Number: 1806294-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR