(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

143397-1G 143397-500MG 143397-5G 143397-25G
77464-892EA 415.09 USD
77464-892 77477-668 77487-790 77498-506
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)trimethylsilane ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₁BrF₄Si
MW: 315.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34836547
CAS Number: 2969341-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR