5-Chloro-1,3-benzodioxole-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chlorobenzo[d][1,3]dioxole-4-carboxylic acid

142305-1G 142305-500MG 142305-5G 142305-25G
77464-884EA 153.47 USD
77464-884 77477-660 77487-778 77498-494
5-Chloro-1,3-benzodioxole-4-carboxylic acid ≥95%
5-Chloro-1,3-benzodioxole-4-carboxylic acid

Precaución: For research use only

Formula: C₈H₅ClO₄
MW: 200.576 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655990
CAS Number: 379229-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR