Pyrene-1-carbonitrile ≥96%

Supplier: AOBChem USA

Warning

141359-1G 141359-500MG 141359-5G
77464-876EA 515 USD
77464-876 77477-652 77487-770
Pyrene-1-carbonitrile ≥96%
Pyrene-1-carbonitrile

Precaución: For research use only

Formula: C₁₇H₉N
MW: 227.2600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127743
CAS Number: 4107-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR