2-Formyl-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

14103-1G 14103-500MG 14103-5G
77464-872EA 539.72 USD
77464-872 77477-648 77487-766
2-Formyl-5-(trifluoromethyl)benzoic acid ≥95%
2-Formyl-5-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₉H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD24479818
CAS Number: 13055-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR