2-Ethoxy-4-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

140371-1G 140371-500MG 140371-5G 140371-25G
77464-868EA 347.11 USD
77464-868 77477-644 77487-762 77498-486
2-Ethoxy-4-methoxybenzoic acid ≥95%
2-Ethoxy-4-methoxybenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017543
CAS Number: 55085-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR