2-(2-Bromo-4-chlorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

14035-1G 14035-5G 14035-25G
77464-864EA 69.01 USD
77464-864 77487-758 77498-484
2-(2-Bromo-4-chlorophenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-4-chlorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₈BrClO₂
MW: 263.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976746
CAS Number: 2221812-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR