1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140293-1G 140293-500MG 140293-5G 140293-25G
77464-860EA 397.58 USD
77464-860 77477-638 77487-754 77498-480
1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787182
CAS Number: 1226420-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR