1-(3-Methoxy-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140236-1G 140236-500MG 140236-5G 140236-25G
77464-856EA 347.11 USD
77464-856 77477-634 77487-750 77498-478
1-(3-Methoxy-4-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Methoxy-4-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34834421
CAS Number: 1600468-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR