1-(3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140209-1G 140209-500MG 140209-5G 140209-25G
77464-852EA 415.09 USD
77464-852 77477-630 77487-746 77498-474
1-(3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD12806389
CAS Number: 1178018-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR