2,2-Dimethyl-1-(4-pentylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140201-1G 140201-500MG 140201-5G 140201-25G
77464-848EA 415.09 USD
77464-848 77477-626 77487-742 77498-470
2,2-Dimethyl-1-(4-pentylphenyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(4-pentylphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₆H₂₆O
MW: 234.377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34834412
CAS Number: 2918955-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR