1-(4-Isopropoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140191-1G 140191-500MG 140191-5G 140191-25G
77464-844EA 415.09 USD
77464-844 77477-622 77487-738 77498-466
1-(4-Isopropoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Isopropoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12787624
CAS Number: 1225711-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR